2-(2,6-Difluorophenyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A134884-250MG A134884-1G
103802-066EA 255 USD
103802-066 77010-086 77298-742
2-(2,6-Difluorophenyl)pyrrolidine 98%
2-(2,6-Difluorophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂N
Boiling Pt: 196.1 °C (760 mm Hg)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09809920
CAS Number: 1016529-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR