(2,6-Dichlorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichlorobenzyl cyanide, 2-(2,6-Dichlorophenyl)acetonitrile

A225356-100G A225356-25G A225356-5G A225356-10G A225356-500G
103850-070EA 6 USD
103850-070 76770-984 77088-762 77088-764 77626-146 77626-148
(2,6-Dichlorophenyl)acetonitrile 98%
(2,6-Dichlorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Boiling Pt: 286.9 °C (760 mm Hg)
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001901
CAS Number: 3215-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR