2-(2,6-Dibromophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A247513-10G A247513-250MG A247513-1G A247513-25G
103811-776EA 55 USD
103811-776 103851-612 77031-412 77031-414 77090-950 77090-952
2-(2,6-Dibromophenyl)acetic acid 97%
2-(2,6-Dibromophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09832981
CAS Number: 901310-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR