2-Maleimidoacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetic acid

A110455-100MG A110455-10G A110455-250MG A110455-25G A110455-5G
103797-666EA 25 USD
103797-666 103823-472 77257-632 77257-634 77257-636 77257-638 77310-266
2-Maleimidoacetic acid 97%
2-Maleimidoacetic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₄
MW: 155.11 g/mol
Boiling Pt: 376.7 °C (760 mm Hg)
Melting Pt: 92…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00457254
CAS Number: 25021-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR