2,4-Dimethoxyphenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A118596-25G A118596-1G A118596-5G
103799-080EA 17 USD
103799-080 77246-014 77297-610 77297-612
2,4-Dimethoxyphenylboronic acid pinacol ester 97%
2,4-Dimethoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₄
MW: 264.13 g/mol
Boiling Pt: 367.9 °C (760 mm Hg)
Flash Pt: 176.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06795688
CAS Number: 214360-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR