2-(2,4-Difluorophenyl)pyridine 98%
Supplier: Ambeed
|
Warning
|
A150452-100G
A150452-10G
A150452-5G
103804-168EA
9
USD
103804-168
103842-302
77235-418
77235-420
77317-820
2-(2,4-Difluorophenyl)pyridine 98%
2-(2,4-Difluorophenyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₁H₇F₂N MW: 191.18 g/mol Melting Pt: 20 °C Flash Pt: 103.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD07787526 CAS Number: 391604-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...