2-(2,4-Dibromophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A180038-100MG A180038-250MG A180038-1G A180038-5G
103807-500EA 110 USD
103807-500 103845-854 77011-958 77011-960 77083-332 77083-334
2-(2,4-Dibromophenyl)acetic acid 97%
2-(2,4-Dibromophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13185526
CAS Number: 98434-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR