2-(2,3-Dichlorophenyl)ethanol 98%

Supplier: Ambeed
Warning

A175405-1G A175405-10G
103807-038EA 12 USD
103807-038 103845-346 77082-688 77248-130
2-(2,3-Dichlorophenyl)ethanol 98%
2-(2,3-Dichlorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
Boiling Pt: 279.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04112742
CAS Number: 114686-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR