(2,3-Dichlorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichlorophenylacetonitrile, 2-(2,3-Dichlorophenyl)acetonitrile, 2,3-Dichlorobenzyl cyanide

A164328-100G A164328-10G A164328-1G
103805-850EA 6 USD
103805-850 103844-110 77096-248 77096-250 77320-270
(2,3-Dichlorophenyl)acetonitrile 98%
(2,3-Dichlorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Boiling Pt: 289.6 °C (760 mm Hg)
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01861392
CAS Number: 3218-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR