(2,3,6-Trifluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A516827-1G A516827-25G A516827-100G A516827-250MG A516827-5G
103828-462EA 29 USD
103828-462 103839-568 76843-556 76843-558 77241-656 77241-658 77307-680
(2,3,6-Trifluorophenyl)acetic acid 97%
(2,3,6-Trifluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₂F₃CH₂COOH
Melting Pt: 111…114 °C
Flash Pt: 104.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00061217
CAS Number: 114152-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR