Phthalimidoacetaldehyde diethyl acetal 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2,2-Diethoxyethyl)isoindoline-1,3-dione, Phthalimidoacetaldehyd Diethylacetal
A359272-100G
A359272-10G
A359272-500G
A359272-25G
A359272-5G
103837-230EA
17
USD
103837-230
76724-558
76724-560
76724-562
76840-394
76840-396
Phthalimidoacetaldehyde diethyl acetal 98%
Phthalimidoacetaldehyde diethyl acetal
Precaución: for Research Use Only
Formula:
C₁₄H₁₇NO₄ Boiling Pt: 372.3 °C (760 mm Hg) Melting Pt: 72…74 °C Flash Pt: 179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005901 CAS Number: 78902-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...