2-(2-(Bromomethyl)phenyl)acetonitrile 97%

Supplier: Ambeed
Danger

A314097-1G A314097-100MG A314097-250MG
103825-024EA 105 USD
103825-024 103835-940 76838-750 77251-020 77251-022
2-(2-(Bromomethyl)phenyl)acetonitrile 97%
2-(2-(Bromomethyl)phenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD00462772
CAS Number: 73217-11-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR