Diethylene glycol monobenzyl ether 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[2-(Benzyloxy)ethoxy]ethanol, 2-(2-(Benzyloxy)ethoxy)ethanol
A240284-10G
A240284-25G
A240284-100G
103809-702EA
198
USD
103809-702
103851-144
77249-754
77304-192
77615-982
Diethylene glycol monobenzyl ether 98%
Diethylene glycol monobenzyl ether
Precaución: for Research Use Only
Formula:
C₁₁H₁₆O₃ MW: 196.25 g/mol Boiling Pt: 311.2 °C (760 mm Hg) Flash Pt: 142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00029649 CAS Number: 2050-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...