[2-(Aminomethyl)phenyl]acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: [2-(Aminomethyl)phenyl]acetic Acid, 2-Aminomethylphenylacetic acid, 2-(2-(Aminomethyl)phenyl)acetic acid

A318564-1G A318564-5G A318564-100MG A318564-10G A318564-250MG
103825-174EA 9 USD
103825-174 103836-108 76838-964 76838-966 77251-094 77251-096 77251-098
[2-(Aminomethyl)phenyl]acetic acid 98%
[2-(Aminomethyl)phenyl]acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 328 °C (760 mm Hg)
Flash Pt: 152.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04038440
CAS Number: 40851-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR