2-(2-(4-Fluorophenyl)-2-oxo-1-phenylethyl)-4-methyl-3-oxo-N-phenylpentanamide 98%
Supplier: Ambeed
|
Warning
|
A234924-100G
A234924-10G
A234924-1G
A234924-500G
A234924-25G
103850-750EA
6
USD
103850-750
77000-276
77000-278
77000-280
77000-282
77089-752
2-(2-(4-Fluorophenyl)-2-oxo-1-phenylethyl)-4-methyl-3-oxo-N-phenylpentanamide 98%
2-(2-(4-Fluorophenyl)-2-oxo-1-phenylethyl)-4-methyl-3-oxo-N-phenylpentanamide
Precaución: for Research Use Only
Formula:
C₂₆H₂₄FNO₃ MW: 417.4700 g/mol Boiling Pt: 631.4 °C (760 mm Hg) Melting Pt: 196…198 °C Flash Pt: 335.645 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04117986 CAS Number: 125971-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...