2-[(4-Chlorophenyl)acetyl]benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(4-Chlorophenyl)acetyl)benzoic acid

A138356-100G A138356-10G A138356-1G A138356-25G A138356-5G
103802-602EA 7 USD
103802-602 76972-450 76972-452 77298-982 77315-622 77315-624
2-[(4-Chlorophenyl)acetyl]benzoic acid 97%
2-[(4-Chlorophenyl)acetyl]benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁ClO₃
MW: 274.7 g/mol
Boiling Pt: 456.8 °C (760 mm Hg)
Flash Pt: 230.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07369702
CAS Number: 53242-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR