m-PEG3-Amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(2-Methoxyethoxy)ethoxy)ethanamine

A132998-250MG A132998-1G A132998-5G A132998-100G A132998-25G
103801-812EA 19 USD
103801-812 76972-190 76972-192 77298-598 77314-404 77670-888 77670-890
m-PEG3-Amine 98%
m-PEG3-Amine

Precaución: for Research Use Only

Formula: C₇H₁₇NO₃
Flash Pt: 86.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17215909
CAS Number: 74654-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR