2-(2-(2-Chloroethoxy)ethoxy)ethanol 97%

Supplier: Ambeed
Warning

A222346-5G A222346-100G
103849-856EA 7 USD
103849-856 77303-626 77568-144
2-(2-(2-Chloroethoxy)ethoxy)ethanol 97%
2-(2-(2-Chloroethoxy)ethoxy)ethanol

Precaución: For research use only.

Formula: C₆H₁₃ClO₃
MW: 168.62 g/mol
Boiling Pt: 247.6 °C (760 mm Hg)
Flash Pt: 107.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002874
CAS Number: 5197-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR