2-[2-(2-Aminoethoxy)ethoxy]ethanol 98%

Supplier: Ambeed
Danger

Synonyms: 2-(2-(2-Aminoethoxy)ethoxy)ethanol

A536745-1G A536745-25G A536745-5G A536745-100MG A536745-10G A536745-250MG
103828-600EA 33 USD
103828-600 103839-722 76843-752 76843-754 76843-756 77253-632 77253-634 77253-636
2-[2-(2-Aminoethoxy)ethoxy]ethanol 98%
2-[2-(2-Aminoethoxy)ethoxy]ethanol

Precaución: for Research Use Only

Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 253.8 °C (760 mm Hg)
Flash Pt: 107.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367495
CAS Number: 6338-55-2
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR