2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Methanesulphonic acid 2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl ester, 2-(2-(2,2,2-Trifluoroethoxy)phenoxy)ethyl methanesulphonate

A101536-10G A101536-250MG A101536-25G A101536-1G A101536-5G
103796-012EA 10 USD
103796-012 103821-696 76869-166 76869-168 76869-170 77282-752 77282-754
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulfonate 98%
2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃O₅S
MW: 314.28 g/mol
Boiling Pt: 396.4 °C (760 mm Hg)
Flash Pt: 193.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16038209
CAS Number: 160969-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR