Amino-PEG4-alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(2-(2-Aminoethoxy)ethoxy)ethoxy)ethanol

A201204-1G A201204-25G A201204-5G A201204-100MG A201204-10G A201204-250MG A201204-100G
103817-278EA 11 USD
103817-278 103847-916 77085-988 77085-990 77085-992 77192-312 77192-314 77192-316 77550-292
Amino-PEG4-alcohol 95%
Amino-PEG4-alcohol

Precaución: for Research Use Only

Formula: C₈H₁₉NO₄
Boiling Pt: 301.6 °C (760 mm Hg)
Flash Pt: 136.2 °C
Storage Temperature: Freezer
MDL Number: MFCD01752874
CAS Number: 86770-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR