2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl)-1H-1,2,4-triazole-3(2H)-thione, Prothioconazole

A320652-100G A320652-10G A320652-250MG A320652-1G A320652-25G A320652-5G
103825-240EA 9 USD
103825-240 103836-174 77167-274 77167-276 77167-278 77306-214 77306-216 77306-218
2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione 97%
2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione

Precaución: for Research Use Only

Formula: C₁₄H₁₅Cl₂N₃OS
MW: 344.259 g/mol
Boiling Pt: 486.7 °C (760 mm Hg)
Flash Pt: 248.2 °C
Storage Temperature: Ambient
MDL Number: MFCD08273847
CAS Number: 178928-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR