Boc(aminophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-((tert-Butoxycarbonyl)amino)phenyl)acetic acid

A492444-1G A492444-5G A492444-10G A492444-250MG
103828-226EA 52 USD
103828-226 103839-302 76843-264 76843-266 76979-794 76979-796
Boc(aminophenyl)acetic acid 97%
Boc(aminophenyl)acetic acid

Precaución: For research use only

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Boiling Pt: 355.6±25.0 °C (760 mm Hg)
Flash Pt: 168.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750482
CAS Number: 135807-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR