2-(2-(((Benzyloxy)carbonyl)amino)thiazol-4-yl)-5-((3-methylbut-2-en-1-yl)oxy)-5-oxopent-2-enoic acid 97%

Supplier: Ambeed
Warning

A141081-100MG A141081-250MG A141081-1G A141081-5G
103802-950EA 17 USD
103802-950 76689-016 76689-018 77299-198 77299-200
2-(2-(((Benzyloxy)carbonyl)amino)thiazol-4-yl)-5-((3-methylbut-2-en-1-yl)oxy)-5-oxopent-2-enoic acid 97%
2-(2-(((Benzyloxy)carbonyl)amino)thiazol-4-yl)-5-((3-methylbut-2-en-1-yl)oxy)-5-oxopent-2-enoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O₆S
MW: 430.47 g/mol
Melting Pt: 192…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750993
CAS Number: 115065-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR