2-(2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)ethoxy)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethoxy)acetic acid, Fmoc-AEA, Fmoc-NH-PEG1-CH2COOH
A166606-250MG
A166606-1G
A166606-5G
A166606-100MG
103806-082EA
54
USD
103806-082
103844-346
77163-360
77320-618
77320-620
77611-624
2-(2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)ethoxy)acetic acid 97%
2-(2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)ethoxy)acetic acid
Precaución: Sealed in dry, at 2…8°C
Formula:
C₁₉H₁₉NO₅ MW: 341.358 g/mol Boiling Pt: 602.6 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06796876 CAS Number: 260367-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...