2-(1H-Pyrazol-1-yl)ethanol 95%

Supplier: Ambeed
Warning

A841323-1G A841323-5G
103841-208EA 138 USD
103841-208 76845-448 76845-450
2-(1H-Pyrazol-1-yl)ethanol 95%
2-(1H-Pyrazol-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₈N₂O
Boiling Pt: 236.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00046076
CAS Number: 6314-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR