2-(1-Phenylethyl)phenol 95%

Supplier: Ambeed
Warning

A160704-100MG A160704-250MG A160704-1G
103805-408EA 608 USD
103805-408 77150-874 77150-876 77300-486
2-(1-Phenylethyl)phenol 95%
2-(1-Phenylethyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
Boiling Pt: 313.9±11.0 °C (760 mm Hg)
Flash Pt: 148.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01707270
CAS Number: 4237-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR