2-(1H-Pyrazol-4-yl)ethanol 98%

Supplier: Ambeed
Warning

A236435-10G A236435-25G A236435-1G
103811-092EA 15 USD
103811-092 103850-864 77013-662 77089-908 77304-050
2-(1H-Pyrazol-4-yl)ethanol 98%
2-(1H-Pyrazol-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₈N₂O
Boiling Pt: 307.6 °C (760 mm Hg)
Melting Pt: 52…56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419343
CAS Number: 180207-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR