2-(1H-Pyrazol-3-yl)acetic acid hydrochloride 97%

Supplier: Ambeed
Warning

A231138-100MG A231138-250MG
103810-754EA 95 USD
103810-754 103850-502 76826-448 77683-674
2-(1H-Pyrazol-3-yl)acetic acid hydrochloride 97%
2-(1H-Pyrazol-3-yl)acetic acid hydrochloride

Precaución: For research use only.

Formula: C₅H₇ClN₂O₂
MW: 162.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22201145
CAS Number: 118054-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR