1H-Indol-4-ylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1H-Indol-4-yl)acetic acid

A212531-100MG A212531-250MG A212531-1G A212531-5G
103818-208EA 95 USD
103818-208 103848-904 76785-722 76785-724 77087-372 77303-248
1H-Indol-4-ylacetic acid 98%
1H-Indol-4-ylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Boiling Pt: 415 °C (760 mm Hg)
Melting Pt: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD09837549
CAS Number: 16176-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR