(1-Indolyl)acetic acid 96%
Supplier: Ambeed
|
Warning
|
A133678-100G
A133678-10G
A133678-250MG
A133678-25G
A133678-1G
A133678-5G
103801-900EA
6
USD
103801-900
77027-192
77027-194
77027-196
77298-648
77314-578
77314-580
(1-Indolyl)acetic acid 96%
(1-Indolyl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂ Boiling Pt: 384.5 °C (760 mm Hg) Melting Pt: 174…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047262 CAS Number: 24297-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...