2-(1H-Indazol-3-yl)acetic acid 97%

Supplier: Ambeed
Warning

A108073-250MG A108073-5G
103797-286EA 30 USD
103797-286 103823-060 77135-784 77309-904
2-(1H-Indazol-3-yl)acetic acid 97%
2-(1H-Indazol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
Boiling Pt: 434.3 °C (760 mm Hg)
Melting Pt: 186…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08236726
CAS Number: 26663-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR