2-(1H-Imidazol-5-yl)acetic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Imidazoleacetic acid hydrochloride, Imidazole-4-acetic acid monohydrochloride

A138638-100G A138638-10G A138638-1G A138638-5G
103802-646EA 47 USD
103802-646 77285-626 77285-628 77315-692 77315-694
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 97%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇ClN₂O₂
MW: 162.58 g/mol
Melting Pt: 218…222 °C
Flash Pt: 224.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00012698
CAS Number: 3251-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR