2-(1H-Imidazol-1-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Imidazoleacetic acid, Imidazol-1-yl-acetic acid, 1-(Carboxymethyl)imidazole, 2-(1-Imidazolyl)acetic acid

A366382-5G A366382-10G A366382-100G A366382-500G
103810-224EA 248 USD
103810-224 103837-414 77306-770 77572-092 77748-726 77748-727
2-(1H-Imidazol-1-yl)acetic acid 97%
2-(1H-Imidazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.1133 g/mol
Boiling Pt: 392.1 °C (760 mm Hg)
Melting Pt: 258 °C
Flash Pt: 190.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01721800
CAS Number: 22884-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR