2-(1H-Benzo[d]imidazol-6-yl)acetic acid 98%

Supplier: Ambeed
Warning

A336668-100MG A336668-250MG A336668-1G
103825-730EA 171 USD
103825-730 103836-702 76989-354 76989-356 77306-502
2-(1H-Benzo[d]imidazol-6-yl)acetic acid 98%
2-(1H-Benzo[d]imidazol-6-yl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690200
CAS Number: 473895-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR