(1-Ethoxyethylidene)malononitrile 99+%

Supplier: Ambeed
Danger

Synonyms: 2-(1-Ethoxyethylidene)malononitrile, 2-(1-Ethoxy-ethylidene)-malononitrile

A229793-100G A229793-10G A229793-1G A229793-25G A229793-5G
103819-496EA 9 USD
103819-496 103850-398 76771-110 76771-112 77089-244 77089-246 77089-248
(1-Ethoxyethylidene)malononitrile 99+%
(1-Ethoxyethylidene)malononitrile

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 312.6 °C (760 mm Hg)
Melting Pt: 90…92 °C
Flash Pt: 137.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001852
CAS Number: 5417-82-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR