1-Fmoc-4-piperidineacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidin-4-yl)acetic acid, Fmoc-4-piperidylacetic acid

A168561-100MG A168561-250MG A168561-25G
103806-296EA 47 USD
103806-296 103844-550 76998-136 77671-380 77671-382
1-Fmoc-4-piperidineacetic acid 97%
1-Fmoc-4-piperidineacetic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₃NO₄
MW: 365.429 g/mol
Boiling Pt: 575 °C (760 mm Hg)
Melting Pt: 135…138 °C
Flash Pt: 301.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273478
CAS Number: 180181-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR