2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[[tert-Butyl(dimethyl)silyl]oxy]ethan-1-ol, 2-((tert-Butyldimethylsilyl)oxy)ethanol
A352441-10G
A352441-5G
A352441-1000G
A352441-500G
103837-070EA
5
USD
103837-070
76828-606
77306-632
77613-010
77613-012
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol 98%
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol
Precaución: for Research Use Only
Formula:
C₈H₂₀O₂Si MW: 176.33 g/mol Boiling Pt: 186.1 °C (760 mm Hg) Melting Pt: <0 °C Flash Pt: 55 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09261150 CAS Number: 102229-10-7 ADR: 3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...