2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((tert-Butyldimethylsilyl)oxy)ethanamine, (2-Aminoethoxy)(tert-butyl)dimethylsilane
A192081-100G
A192081-10G
A192081-1G
A192081-25G
A192081-5G
103816-416EA
5
USD
103816-416
103847-020
76825-298
76825-300
77084-892
77084-894
77084-896
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine 95%
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine
Precaución: for Research Use Only
Formula:
C₈H₂₁NOSi MW: 175.35 g/mol Boiling Pt: 178.4±13.0 °C (760 mm Hg) Flash Pt: 61.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18206132 CAS Number: 101711-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...