2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Ser-OH

A105643-100G A105643-10G A105643-25G A105643-1G A105643-5G
103796-916EA 6 USD
103796-916 103822-652 76970-416 76970-418 76970-420 77309-522 77309-524
2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid 95%
2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₅
MW: 205.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037243
CAS Number: 3850-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR