Boc-2-Cyclohexylglycine 97%

Supplier: Ambeed
Warning

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid

A102149-100MG A102149-10G A102149-25G A102149-1G A102149-5G
103796-318EA 8 USD
103796-318 103822-012 77107-226 77107-228 77107-230 77308-886 77308-888
Boc-2-Cyclohexylglycine 97%
Boc-2-Cyclohexylglycine

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.326 g/mol
Boiling Pt: 407.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04115511
CAS Number: 35264-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR