2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 2-((6-Chloro-2-methylpyrimidin-4-yl)amino)-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide, N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide

A160437-100G A160437-10G A160437-250MG A160437-1G A160437-25G A160437-5G
103805-378EA 6 USD
103805-378 103843-606 77028-626 77028-628 77028-630 77319-540 77319-542 77319-544
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide 95%
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₆H₁₃Cl₂N₅OS
MW: 394.27 g/mol
Melting Pt: 333…335 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11045075
CAS Number: 302964-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR