2-((5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-2-yl)oxy)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Ramage Linker, Fmoc-Suberol

A286441-1G A286441-5G A286441-10G
103813-708EA 15 USD
103813-708 103834-844 76837-378 76837-380 77002-264
2-((5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-2-yl)oxy)acetic acid 97%
2-((5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-2-yl)oxy)acetic acid

Precaución: for Research Use Only

Formula: C₃₂H₂₇NO₅
MW: 505.6 g/mol
Boiling Pt: 724.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07783960
CAS Number: 212783-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR