2-((4-Methoxybenzyl)oxy)acetic acid 95%

Supplier: Ambeed
Warning

A233656-1G A233656-25G A233656-5G A233656-100MG A233656-250MG
103810-900EA 14 USD
103810-900 103850-658 77089-618 77089-620 77089-622 77113-076 77113-078
2-((4-Methoxybenzyl)oxy)acetic acid 95%
2-((4-Methoxybenzyl)oxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
Boiling Pt: 367.4 °C (760 mm Hg)
Flash Pt: 146.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861881
CAS Number: 88920-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR