XRK3F2 99+%

Supplier: Ambeed
Warning

Synonyms: XRK3F2 HCl, 2-((3,4-Bis((4-fluorobenzyl)oxy)benzyl)amino)ethan-1-ol hydrochloride

A878726-10MG A878726-1MG A878726-25MG A878726-50MG A878726-5MG
103791-736EA 29 USD
103791-736 103799-738 76654-816 76654-818 76654-820 76845-576 76845-578
XRK3F2 99+%
XRK3F2

Precaución: for Research Use Only

Formula: C₂₃H₂₄ClF₂NO₃
MW: 435.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812777
CAS Number: 2375193-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR