2-Chloro-5-fluoro-4-(2-hydroxyethylamino)pyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-((2-Chloro-5-fluoropyrimidin-4-yl)amino)ethanol

A287972-1G A287972-250MG
103813-778EA 44 USD
103813-778 103834-920 76837-472 77194-246
2-Chloro-5-fluoro-4-(2-hydroxyethylamino)pyrimidine 97%
2-Chloro-5-fluoro-4-(2-hydroxyethylamino)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClFN₃O
MW: 191.591 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02629720
CAS Number: 16255-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR