FABP4 inhibitor 98%

Supplier: Ambeed
Warning

Synonyms: BMS-309403, 2-((2'-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)-[1,1'-biphenyl]-3-yl)oxy)acetic acid

A115684-1MG A115684-5MG
103791-432EA 96 USD
103791-432 103792-040 103792-042 103799-292 77233-890 77311-280
FABP4 inhibitor 98%
FABP4 inhibitor

Precaución: for Research Use Only

Formula: C₃₁H₂₆N₂O₃
MW: 474.5500 g/mol
Boiling Pt: 657.5±55.0 °C (760 mm Hg)
Flash Pt: 351.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09991687
CAS Number: 300657-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR