2-((1-Benzylpiperidin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one 98%

Supplier: Ambeed
Warning

Synonyms: Donepezil

A127504-100G A127504-10G A127504-1G A127504-25G A127504-5G
103800-968EA 14 USD
103800-968 77108-996 77108-998 77298-220 77313-206 77313-208
2-((1-Benzylpiperidin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one 98%
2-((1-Benzylpiperidin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₂₄H₂₉NO₃
Boiling Pt: 527.9 °C (760 mm Hg)
Melting Pt: 207 °C
Flash Pt: 273.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00912833
CAS Number: 120014-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR