1-Tritylimidazole-4-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Trityl-1H-imidazole-4-carboxaldehyde, 1-Trityl-1H-imidazole-4-carbaldehyde, 1-Tritylimidazole-4-carboxaldehyde
A125745-100G
A125745-1G
A125745-10G
103800-716EA
10
USD
103800-716
76782-002
77312-866
77599-820
1-Tritylimidazole-4-carbaldehyde 97%
1-Tritylimidazole-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₂₃H₁₈N₂O MW: 338.41 g/mol Boiling Pt: 502.8 °C (760 mm Hg) Melting Pt: 180…190 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179554 CAS Number: 33016-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...