1-Tritylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: Clotrimazole Impurity F, 1-Trityl-1H-imidazole

A181599-100G A181599-10G A181599-250MG A181599-1G A181599-25G A181599-5G
103807-628EA 7 USD
103807-628 103846-004 77096-878 77096-880 77096-882 77301-732 77301-734 77301-736
1-Tritylimidazole 98%
1-Tritylimidazole

Precaución: for Research Use Only

Formula: C₂₂H₁₈N₂
Boiling Pt: 451.3 °C (760 mm Hg)
Melting Pt: 220…224 °C
Flash Pt: 226.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00229427
CAS Number: 15469-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR